Assay Details
Gene Symbol: LOC4342175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015789084.2 | XP_015644570.1 | 3 - 4 | 957 | 76 | |
GenBank mRNA | AK101907.1 | - | 3 - 4 | 958 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
