Assay Details
Gene Symbol: LOC4342371 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791239.2 | XP_015646725.1 | 1 - 2 | 241 | 60 | |
GenBank mRNA | AK068893.1 | - | 1 - 2 | 227 | 60 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015791239.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
