Assay Details
Gene Symbol: LOC4342635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791930.1 | XP_015647416.1 | 2 - 3 | 515 | 85 | |
XM_015791931.1 | XP_015647417.1 | 2 - 3 | 515 | 85 | ||
GenBank mRNA | AK070547.1 | - | 1 - 2 | 53 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
