Assay Details
Gene Symbol: LOC4342635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791930.1 | XP_015647416.1 | 8 - 9 | 1105 | 99 | |
XM_015791931.1 | XP_015647417.1 | 6 - 7 | 820 | 99 | ||
GenBank mRNA | AK070547.1 | - | 7 - 8 | 643 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
