Assay Details
Gene Symbol: LOC4343634 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015790018.2 | XP_015645504.1 | 5 - 6 | 634 | 105 | |
GenBank mRNA | AK068245.1 | - | 3 - 4 | 550 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
