Assay Details
| Gene Symbol: LOC4343835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792400.2 | XP_015647886.1 | 1 - 2 | 136 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015792400.2 transcripts.
Set Membership: |
|

LOC4343835
Genomic Map