Assay Details
| Gene Symbol: LOC4343835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792400.2 | XP_015647886.1 | 7 - 8 | 1205 | 61 | |
| GenBank mRNA | AK101352.1 | - | 7 - 8 | 1202 | 61 | |
More Information
Set Membership: |
|

LOC4343835
Genomic Map