Assay Details
| Gene Symbol: LOC4343883 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789156.2 | XP_015644642.1 | 5 - 6 | 1611 | 97 | |
| GenBank mRNA | AK242420.1 | - | 1 - 2 | 180 | 97 | |
More Information
Set Membership: |
|

LOC4343883
Genomic Map