Assay Details
| Gene Symbol: LOC4343936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790045.2 | XP_015645531.1 | 1 - 2 | 618 | 126 | |
| GenBank mRNA | AK063671.1 | - | 1 - 2 | 618 | 126 | |
More Information
Set Membership: |
|

LOC4343936
Genomic Map