Assay Details
| Gene Symbol: LOC4344071 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791886.1 | XP_015647372.1 | 1 - 2 | 213 | 59 | |
| GenBank mRNA | AK119728.1 | - | 1 - 2 | 178 | 59 | |
| AK122129.1 | - | 1 - 2 | 179 | 59 | ||
| BT129187.1 | - | 1 - 2 | 181 | 59 | ||
| BT130576.1 | - | 1 - 2 | 172 | 59 | ||
| BT131414.1 | - | 1 - 2 | 170 | 59 | ||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length less than or equal to 70  Recommended Assay  Oryza sativa  Probe spans exons | 
Panther Classification:
 Back To Top
 Back To Top
 LOC4344071
LOC4344071 Genomic Map
Genomic Map Back To Top
 Back To Top