Assay Details
Gene Symbol: LOC4344564 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794644.2 | XP_015650130.1 | 5 - 6 | 541 | 107 | |
GenBank mRNA | AK071097.1 | - | 5 - 6 | 429 | 107 | |
AK103777.1 | - | 5 - 6 | 396 | 107 | ||
EF122472.1 | - | 5 - 6 | 414 | 107 | ||
GQ848031.1 | - | 5 - 6 | 412 | 107 | ||
JF969200.1 | - | 5 - 6 | 392 | 107 | ||
JN848809.1 | - | 4 - 5 | 352 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
