Assay Details
| Gene Symbol: LOC4344564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794644.2 | XP_015650130.1 | 8 - 9 | 836 | 79 | |
| GenBank mRNA | AK071097.1 | - | 8 - 9 | 724 | 79 | |
| AK103777.1 | - | 8 - 9 | 691 | 79 | ||
| EF122472.1 | - | 8 - 9 | 709 | 79 | ||
| GQ848031.1 | - | 8 - 9 | 707 | 79 | ||
| JF969200.1 | - | 8 - 9 | 687 | 79 | ||
| JN848809.1 | - | 7 - 8 | 647 | 79 | ||
More Information
Set Membership: |
|

LOC4344564
Genomic Map