Assay Details
Gene Symbol: LOC9266786 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792644.2 | XP_015648130.1 | 8 - 9 | 957 | 82 | |
XM_015792645.2 | XP_015648131.1 | 8 - 9 | 714 | 82 | ||
GenBank mRNA | AK058577.1 | - | 398 | 82 | ||
AK242866.1 | - | 2203 | 82 |
Gene Symbol: LOC4323897 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK288851.1 | - | 549 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
