Assay Details
| Gene Symbol: LOC4344690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794641.2 | XP_015650127.1 | 2 - 3 | 1054 | 83 | |
| GenBank mRNA | AK070077.1 | - | 2 - 3 | 714 | 83 | |
More Information
Set Membership: |
|

LOC4344690
Genomic Map