Assay Details
Gene Symbol: LOC4344777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795218.1 | XP_015650704.1 | 1 - 2 | 317 | 144 | |
GenBank mRNA | AK101266.1 | - | 1 - 2 | 318 | 144 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015795218.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
