Assay Details
Gene Symbol: LOC4344777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795218.1 | XP_015650704.1 | 2 - 3 | 738 | 123 | |
GenBank mRNA | AK101266.1 | - | 2 - 3 | 737 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
