Assay Details
Gene Symbol: LOC9269022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765644.2 | XP_015621130.1 | 3 - 4 | 1154 | 81 | |
XM_015765646.2 | XP_015621132.1 | 4 - 5 | 1068 | 81 | ||
XM_015765647.2 | XP_015621133.1 | 3 - 4 | 1153 | 81 |
Gene Symbol: LOC4344777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795218.1 | XP_015650704.1 | 3 - 4 | 81 | ||
GenBank mRNA | AK101266.1 | - | 3 - 4 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
