Assay Details
Gene Symbol: LOC4344777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795218.1 | XP_015650704.1 | 4 - 5 | 892 | 82 | |
GenBank mRNA | AK101266.1 | - | 4 - 5 | 891 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
