Assay Details
Gene Symbol: LOC9269022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765644.2 | XP_015621130.1 | 5 - 6 | 1318 | 59 | |
XM_015765646.2 | XP_015621132.1 | 6 - 7 | 1232 | 59 | ||
XM_015765647.2 | XP_015621133.1 | 5 - 6 | 1317 | 59 |
Gene Symbol: LOC4344777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795218.1 | XP_015650704.1 | 5 - 6 | 965 | 59 | |
GenBank mRNA | AK101266.1 | - | 5 - 6 | 964 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
