Assay Details
Gene Symbol: LOC4345209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795406.2 | XP_015650892.1 | 2 - 3 | 328 | 60 | |
GenBank mRNA | AK061379.1 | - | 1 - 2 | 137 | 60 | |
AK099389.1 | - | 1 - 2 | 103 | 60 | ||
AK101640.1 | - | 1 - 2 | 103 | 60 |
Gene Symbol: LOC107277955 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015783096.1 | XP_015638582.1 | 2 - 3 | 368 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
