Assay Details
Gene Symbol: LOC4345209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795406.2 | XP_015650892.1 | 4 - 5 | 483 | 72 | |
GenBank mRNA | AK061379.1 | - | 3 - 4 | 292 | 72 | |
AK099389.1 | - | 3 - 4 | 258 | 72 | ||
AK101640.1 | - | 3 - 4 | 258 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
