Assay Details
Gene Symbol: LOC4345209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795406.2 | XP_015650892.1 | 5 - 6 | 585 | 130 | |
GenBank mRNA | AK061379.1 | - | 4 - 5 | 394 | 130 | |
AK099389.1 | - | 4 - 5 | 360 | 130 | ||
AK101640.1 | - | 4 - 5 | 360 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
