Assay Details
Gene Symbol: LOC4345410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792929.2 | XP_015648415.1 | 1 - 2 | 275 | 78 | |
GenBank mRNA | AK103189.1 | - | 1 - 2 | 265 | 78 | |
BT129392.1 | - | 1 - 2 | 164 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
