Assay Details
| Gene Symbol: LOC4346376 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756936.2 | XP_015612422.1 | 4 - 5 | 470 | 97 | |
| XM_015756937.2 | XP_015612423.1 | 4 - 5 | 475 | 97 | ||
| GenBank mRNA | AK241825.1 | - | 4 - 5 | 430 | 97 | |
| KJ472480.1 | - | 4 - 5 | 393 | 97 | ||
| KJ472481.1 | - | 3 - 4 | 312 | 97 | ||
| KJ472482.1 | - | 3 - 4 | 312 | 97 | ||
More Information
Set Membership: |
|

LOC4346376
Genomic Map