Assay Details
| Gene Symbol: LOC4346376 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756936.2 | XP_015612422.1 | 5 - 6 | 575 | 151 | |
| XM_015756937.2 | XP_015612423.1 | 5 - 6 | 580 | 151 | ||
| GenBank mRNA | AK241825.1 | - | 5 - 6 | 535 | 151 | |
| KJ472480.1 | - | 5 - 6 | 498 | 151 | ||
| KJ472481.1 | - | 4 - 5 | 417 | 151 | ||
| KJ472482.1 | - | 4 - 5 | 417 | 151 | ||
More Information
Set Membership: |
|

LOC4346376
Genomic Map