Assay Details
| Gene Symbol: LOC4346376 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756936.2 | XP_015612422.1 | 7 - 8 | 733 | 107 | |
| XM_015756937.2 | XP_015612423.1 | 7 - 8 | 738 | 107 | ||
| GenBank mRNA | KJ472480.1 | - | 7 - 8 | 656 | 107 | |
| KJ472482.1 | - | 6 - 7 | 575 | 107 | ||
More Information
Set Membership: |
|

LOC4346376
Genomic Map