Assay Details
Gene Symbol: LOC4346585 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756599.2 | XP_015612085.1 | 2 - 3 | 254 | 68 | |
XM_015756601.2 | XP_015612087.1 | 2 - 3 | 254 | 68 | ||
GenBank mRNA | AK111610.1 | - | 1 - 2 | 321 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015756599.2, XM_015756601.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
