Assay Details
| Gene Symbol: LOC4347313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755212.2 | XP_015610698.1 | 4 - 5 | 961 | 63 | |
| GenBank mRNA | AB080248.1 | - | 4 - 5 | 967 | 63 | |
| AK063940.1 | - | 4 - 5 | 732 | 63 | ||
| AK102698.1 | - | 2 - 3 | 340 | 63 | ||
More Information
Set Membership: |
|

LOC4347313
Genomic Map