Assay Details
| Gene Symbol: LOC4347552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756893.1 | XP_015612379.1 | 2 - 3 | 471 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4347552| Gene Symbol: LOC4347552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756893.1 | XP_015612379.1 | 2 - 3 | 471 | 96 | |
Set Membership: |
|