Assay Details
Gene Symbol: LOC4347645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015755344.2 | XP_015610830.1 | 10 - 11 | 1341 | 97 | |
XM_015755345.2 | XP_015610831.1 | 10 - 11 | 1362 | 97 | ||
GenBank mRNA | AK072633.1 | - | 10 - 11 | 1260 | 97 | |
AK100918.1 | - | 10 - 11 | 1363 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
