Assay Details
Gene Symbol: LOC4347645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015755344.2 | XP_015610830.1 | 12 - 13 | 2213 | 57 | |
XM_015755345.2 | XP_015610831.1 | 12 - 13 | 2234 | 57 | ||
GenBank mRNA | AK060588.1 | - | 2 - 3 | 784 | 57 | |
AK072633.1 | - | 12 - 13 | 2132 | 57 | ||
AK100918.1 | - | 12 - 13 | 2235 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
