Assay Details
Gene Symbol: LOC4347645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015755344.2 | XP_015610830.1 | 1 - 2 | 142 | 123 | |
XM_015755345.2 | XP_015610831.1 | 1 - 2 | 187 | 123 | ||
GenBank mRNA | AK072633.1 | - | 1 - 2 | 85 | 123 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015755344.2, XM_015755345.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
