Assay Details
Gene Symbol: LOC4347645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015755344.2 | XP_015610830.1 | 2 - 3 | 335 | 66 | |
XM_015755345.2 | XP_015610831.1 | 2 - 3 | 380 | 66 | ||
GenBank mRNA | AK072633.1 | - | 2 - 3 | 278 | 66 | |
AK100918.1 | - | 2 - 3 | 381 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
