Assay Details
Gene Symbol: LOC4347645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015755344.2 | XP_015610830.1 | 4 - 5 | 525 | 86 | |
XM_015755345.2 | XP_015610831.1 | 4 - 5 | 570 | 86 | ||
GenBank mRNA | AK072633.1 | - | 4 - 5 | 468 | 86 | |
AK100918.1 | - | 4 - 5 | 571 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
