Assay Details
Gene Symbol: LOC4347645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015755345.2 | XP_015610831.1 | 8 - 9 | 897 | 64 | |
GenBank mRNA | AK072633.1 | - | 8 - 9 | 795 | 64 | |
AK100918.1 | - | 8 - 9 | 898 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
