Assay Details
Gene Symbol: LOC9268167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015755793.2 | XP_015611279.1 | 1 - 2 | 447 | 98 | |
GenBank mRNA | AK068277.1 | - | 1 - 2 | 453 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
