Assay Details
Gene Symbol: LOC9268167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015755793.2 | XP_015611279.1 | 4 - 5 | 910 | 137 | |
GenBank mRNA | AK068277.1 | - | 3 - 4 | 1124 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
