Assay Details
| Gene Symbol: LOC4348519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758720.2 | XP_015614206.1 | 1 - 2 | 720 | 68 | |
| GenBank mRNA | AK288034.1 | - | 1 - 2 | 458 | 68 | |
| JN604886.1 | - | 1 - 2 | 470 | 68 | ||
More Information
Set Membership: |
|

LOC4348519
Genomic Map