Assay Details
| Gene Symbol: LOC4348716 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757785.2 | XP_015613271.1 | 1 - 2 | 185 | 66 | |
| XM_015757786.2 | XP_015613272.1 | 1 - 2 | 185 | 66 | ||
| GenBank mRNA | AK106662.1 | - | 1 - 2 | 141 | 66 | |
More Information
Set Membership: |
|

LOC4348716
Genomic Map