Assay Details
| Gene Symbol: LOC4349093 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757280.2 | XP_015612766.1 | 9 - 10 | 1164 | 94 | |
| GenBank mRNA | AK073197.1 | - | 9 - 10 | 1170 | 94 | |
More Information
Set Membership: |
|

LOC4349093
Genomic Map