Assay Details
| Gene Symbol: LOC4326136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765825.2 | XP_015621311.1 | 1 - 2 | 250 | 66 | |
| GenBank mRNA | AK101017.1 | - | 1 - 2 | 245 | 66 | |
| AK103194.1 | - | 1 - 2 | 240 | 66 | ||
More Information
Set Membership: |
|

LOC4326136
Genomic Map