Assay Details
| Gene Symbol: LOC4326917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765391.2 | XP_015620877.1 | 1 - 2 | 160 | 64 | |
| GenBank mRNA | AK065323.1 | - | 1 - 2 | 80 | 64 | |
| AK122127.1 | - | 1 - 2 | 80 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015765391.2 transcripts.
Set Membership: |
|

LOC4326917
Genomic Map