Assay Details
Gene Symbol: LOC4325781 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794140.2 | XP_015649626.1 | 1 - 2 | 398 | 106 | |
GenBank mRNA | AK066192.1 | - | 1 - 2 | 393 | 106 | |
AK073246.1 | - | 1 - 2 | 387 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
