Assay Details
| Gene Symbol: LOC4325444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765667.2 | XP_015621153.1 | 3 - 4 | 696 | 150 | |
| GenBank mRNA | AK073323.1 | - | 3 - 4 | 582 | 150 | |
| AK121501.1 | - | 3 - 4 | 703 | 150 | ||
More Information
Set Membership: |
|

LOC4325444
Genomic Map