Assay Details
| Gene Symbol: LOC4324038 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769655.2 | XP_015625141.1 | 5 - 6 | 860 | 72 | |
| GenBank mRNA | AK102942.1 | - | 4 - 5 | 931 | 72 | |
More Information
Set Membership: |
|

LOC4324038
Genomic Map