Assay Details
Gene Symbol: LOC4323939 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766928.2 | 3 - 4 | 651 | 126 | ||
GenBank mRNA | AK061861.1 | - | 3 - 4 | 646 | 126 | |
AK099005.1 | - | 3 - 4 | 606 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
