Assay Details
| Gene Symbol: LOC4326310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766170.2 | XP_015621656.1 | 2 - 3 | 257 | 96 | |
| GenBank mRNA | AK072509.1 | - | 2 - 3 | 254 | 96 | |
More Information
Set Membership: |
|

LOC4326310
Genomic Map