Assay Details
| Gene Symbol: LOC4324449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757571.2 | XP_015613057.1 | 2 - 3 | 707 | 149 | |
| GenBank mRNA | AJ495788.1 | - | 1 - 2 | 245 | 149 | |
| AK103040.1 | - | 2 - 3 | 713 | 149 | ||
More Information
Set Membership: |
|

LOC4324449
Genomic Map