Assay Details
Gene Symbol: LOC4327164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795005.2 | XP_015650491.1 | 4 - 5 | 1086 | 92 | |
GenBank mRNA | AK101948.1 | - | 4 - 5 | 1087 | 92 | |
AK106555.1 | - | 4 - 5 | 909 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
