Assay Details
| Gene Symbol: LOC4324146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767058.2 | XP_015622544.1 | 3 - 4 | 811 | 77 | |
| GenBank mRNA | AK240766.1 | - | 3 - 4 | 654 | 77 | |
More Information
Set Membership: |
|

LOC4324146
Genomic Map